Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGUAGUGAUGUUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ornithorhynchus anatinus | oan-miR-499-5p | MIMAT0006949 |
Salmo salar | ssa-miR-499a-5p | MIMAT0032659 |
Stem-loop Details
Ornithorhynchus anatinus
Salmo salar

More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-499 (v18)
