Assay Details
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | UAUUGCACAUUCACCAGCCUGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila erecta | der-miR-311c-3p | MIMAT0008522 |
Stem-loop Details
Drosophila erecta

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | der-miR-311c-3p |
---|---|
miRBase Accession Number: | MI0009062 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UAUUGCACAUUCACCAGCCUGA |
Species: | Drosophila erecta |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | UAUUGCACAUUCACCAGCCUGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila erecta | der-miR-311c-3p | MIMAT0008522 |