Assay Details
Gene Family ID: | MIPF0000564, mir-1421 |
Mature miRNA Sequence: | UGCAUCGCGUGUGACCUCCCUAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ornithorhynchus anatinus | oan-miR-1421aa-3p | MIMAT0007250 |
Ornithorhynchus anatinus | oan-miR-1421z-3p | MIMAT0007246 |
Stem-loop Details
Ornithorhynchus anatinus

More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-1421z* (v18)
Ochrobactrum anthropi: oan-miR-1421aa* (v18)
Ochrobactrum anthropi: oan-miR-1421aa* (v18)
