Assay Details
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | UAUUGCACAUUCCCCAGCCUGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila ananassae | dan-miR-311b | MIMAT0008475 |
Stem-loop Details
Drosophila ananassae
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dan-miR-311b |
|---|---|
| miRBase Accession Number: | MI0009013 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUUGCACAUUCCCCAGCCUGG |
| Species: | Drosophila ananassae |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | UAUUGCACAUUCCCCAGCCUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila ananassae | dan-miR-311b | MIMAT0008475 |