Assay Details
Gene Family ID: | MIPF0000574, mir-1420 | 
Mature miRNA Sequence: | AUCCAAUCUGGGAGCACUCU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1420b-5p | MIMAT0006924 | 
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
						
							Ochrobactrum anthropi: oan-miR-1420b* (v18)
						
					
				
Genomic Map