Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | AGAUAUGUUUGAUAUUCUUGGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-190-5p | MIMAT0009153 |
Tetranychus urticae | tur-miR-190-5p | MIMAT0023084 |
Stem-loop Details
Bombyx mori
Tetranychus urticae
