Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | GUGACAUCACAUAUACGGCGGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dog | cfa-miR-489 | MIMAT0009860 |
Horse | eca-miR-489 | MIMAT0012943 |
Stem-loop Details
Dog
Horse
