Assay Details
Gene Family ID: | MIPF0001063, MIR1432 |
Mature miRNA Sequence: | CUCAGGAGAGAUGACACCGAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Sorghum bicolor | sbi-miR1432 | MIMAT0011390 |
Zea mays | zma-miR1432-5p | MIMAT0014032 |
Stem-loop Details
Sorghum bicolor
Zea mays
