Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CACACACUGCAAUUACUUUUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rhesus monkey | mml-miR-603 | MIMAT0012781 |
Stem-loop Details
Rhesus monkey

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mml-miR-603 |
---|---|
miRBase Accession Number: | MI0012538 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CACACACUGCAAUUACUUUUUC |
Species: | Rhesus monkey |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CACACACUGCAAUUACUUUUUC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rhesus monkey | mml-miR-603 | MIMAT0012781 |