Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GAAGUUGUUCGUGGUGGAUUCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pongo pygmaeus | ppy-miR-382 | MIMAT0015872 |
Stem-loop Details
Pongo pygmaeus

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ppy-miR-382 |
---|---|
miRBase Accession Number: | MI0014930 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GAAGUUGUUCGUGGUGGAUUCC |
Species: | Pongo pygmaeus |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GAAGUUGUUCGUGGUGGAUUCC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pongo pygmaeus | ppy-miR-382 | MIMAT0015872 |