Assay Details
Gene Family ID: | MIPF0000828, mir-2780 | 
Mature miRNA Sequence: | AAAGGUGUUAAAUGAAGACUUAG | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Bombyx mori | bmo-miR-2780d | MIMAT0013765 | 
Stem-loop Details
Bombyx mori
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bmo-miR-2780d | 
|---|---|
| miRBase Accession Number: | MI0012462 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | AAAGGUGUUAAAUGAAGACUUAG | 
| Species: | Bombyx mori | 
| Product Type: | mirVana® miRNA mimic | 
Gene Family ID: | MIPF0000828, mir-2780 | 
Mature miRNA Sequence: | AAAGGUGUUAAAUGAAGACUUAG | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Bombyx mori | bmo-miR-2780d | MIMAT0013765 |