Assay Details
Gene Family ID: | MIPF0001009, mir-1822 |
Mature miRNA Sequence: | AGUUUCUCUGGGAAAGCUAUCGGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-1822-5p | MIMAT0015266 |
Stem-loop Details
Caenorhabditis elegans
More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-1822* (v17)

Genomic Map