Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAUGUAAUAUGGUCCACUCUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pongo pygmaeus | ppy-miR-380 | MIMAT0015870 |
Stem-loop Details
Pongo pygmaeus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ppy-miR-380 |
|---|---|
| miRBase Accession Number: | MI0014928 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUGUAAUAUGGUCCACUCUU |
| Species: | Pongo pygmaeus |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAUGUAAUAUGGUCCACUCUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pongo pygmaeus | ppy-miR-380 | MIMAT0015870 |