Assay Details
Gene Family ID: | MIPF0000826, mir-2807 |
Mature miRNA Sequence: | CAGCUGCAUACAAAAUUGGGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bombyx mori | bmo-miR-2807d | MIMAT0015433 |
Stem-loop Details
Bombyx mori
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bmo-miR-2807d |
|---|---|
| miRBase Accession Number: | MI0014306 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGCUGCAUACAAAAUUGGGC |
| Species: | Bombyx mori |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000826, mir-2807 |
Mature miRNA Sequence: | CAGCUGCAUACAAAAUUGGGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bombyx mori | bmo-miR-2807d | MIMAT0015433 |