Assay Details
Gene Family ID: | MIPF0000421, mir-879 |
Mature miRNA Sequence: | CUUAUGGCUUCAAGCUUUCGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-879-3p | MIMAT0017291 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-879-3p |
|---|---|
| miRBase Accession Number: | MI0006121 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUAUGGCUUCAAGCUUUCGG |
| Species: | Rat |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000421, mir-879 |
Mature miRNA Sequence: | CUUAUGGCUUCAAGCUUUCGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-879-3p | MIMAT0017291 |