Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | UGGUCGUAUGUAUGACGUCAUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-489-5p | MIMAT0038295 |
| Anolis carolinensis | aca-miR-489-5p | MIMAT0021974 |
| Chicken | gga-miR-489-5p | MIMAT0026632 |
| Chrysemys picta | cpi-miR-489-5p | MIMAT0037897 |
| Columba livia | cli-miR-489-5p | MIMAT0038656 |
| Salmo salar | ssa-miR-489-5p | MIMAT0032657 |
| View More Rows | ||
| Taeniopygia guttata | tgu-miR-489-5p | MIMAT0027024 |
Stem-loop Details
Alligator mississippiensis
Anolis carolinensis
Chicken
Chrysemys picta
Columba livia
Salmo salar
Taeniopygia guttata

Genomic Map