Assay Details
Gene Family ID: | MIPF0000635, mir-1329 |
Mature miRNA Sequence: | CCUCGUAGCUUGAUCACCAUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-1329-3p | MIMAT0021748 |
Stem-loop Details
Anolis carolinensis
More Information
Related Products
miRBase Alias:
Aeromonas caviae Ae398: aca-miR-1329* (v18)

Genomic Map