Assay Details
Gene Family ID: | MIPF0000747, mir-2284 |
Mature miRNA Sequence: | AAAUGUUCGCUUGGCUUUUUCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2284ac | MIMAT0030447 |
Stem-loop Details
Bovine

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bta-miR-2284ac |
---|---|
miRBase Accession Number: | MI0025542 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AAAUGUUCGCUUGGCUUUUUCC |
Species: | Bovine |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000747, mir-2284 |
Mature miRNA Sequence: | AAAUGUUCGCUUGGCUUUUUCC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2284ac | MIMAT0030447 |