Assay Details
Mature miRNA Sequence: | UGAGAUUCUUACAUCUUCAUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Panagrellus redivivus | prd-miR-1175-3p | MIMAT0030503 |
Stem-loop Details
Panagrellus redivivus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | prd-miR-1175-3p |
|---|---|
| miRBase Accession Number: | MI0025572 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGAUUCUUACAUCUUCAUUU |
| Species: | Panagrellus redivivus |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UGAGAUUCUUACAUCUUCAUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Panagrellus redivivus | prd-miR-1175-3p | MIMAT0030503 |