Assay Details
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | UCCUGUACUGAGCUGCCCCGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-486 | MIMAT0011132 |
Stem-loop Details
Dog
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cfa-miR-486 |
|---|---|
| miRBase Accession Number: | MI0008027 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCUGUACUGAGCUGCCCCGA |
| Species: | Dog |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | UCCUGUACUGAGCUGCCCCGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-486 | MIMAT0011132 |