Assay Details
Gene Family ID: | MIPF0000113, mir-188 |
Mature miRNA Sequence: | CAUCCCUUGCAUGGUGGAGGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-188 | MIMAT0009249 |
| Dog | cfa-miR-188 | MIMAT0009880 |
| Eptesicus fuscus | efu-miR-188 | MIMAT0035151 |
| Macaca nemestrina | mne-miR-188 | MIMAT0002310 |
| Pan paniscus | ppa-miR-188 | MIMAT0002311 |
| Pan troglodytes | ptr-miR-188 | MIMAT0002308 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-188 | MIMAT0002309 |
| Rhesus monkey | mml-miR-188-5p | MIMAT0002307 |
| Tupaia chinensis | tch-miR-188-5p | MIMAT0036491 |
Stem-loop Details
Bovine
Dog
Eptesicus fuscus
Macaca nemestrina
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis

Genomic Map