Assay Details
Gene Family ID: | MIPF0000147, mir-325 |
Mature miRNA Sequence: | CCUAGUAGGUGUUCAGUAAGUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-325 | MIMAT0009897 |
| Pig | ssc-miR-325 | MIMAT0002150 |
Stem-loop Details
Dog
Pig
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cfa-miR-325 |
|---|---|
| miRBase Accession Number: | MI0010396 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUAGUAGGUGUUCAGUAAGUGU |
| Species: | Dog, Pig |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000147, mir-325 |
Mature miRNA Sequence: | CCUAGUAGGUGUUCAGUAAGUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-325 | MIMAT0009897 |
| Pig | ssc-miR-325 | MIMAT0002150 |