Assay Details
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | UAAGGUGCAUCUAGUGCAGUUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-18b | MIMAT0003517 |
| Chicken | gga-miR-18b-5p | MIMAT0001141 |
| Chrysemys picta | cpi-miR-18b-5p | MIMAT0037640 |
| Dog | cfa-miR-18b | MIMAT0009831 |
| Taeniopygia guttata | tgu-miR-18b | MIMAT0014607 |
Stem-loop Details
Bovine
Chicken
Chrysemys picta
Dog
Taeniopygia guttata

Genomic Map