Assay Details
Gene Family ID: | MIPF0000117, mir-139 |
Mature miRNA Sequence: | UCUACAGUGCACGUGUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Lagothrix lagotricha | lla-miR-139 | MIMAT0002472 |
| Pan paniscus | ppa-miR-139 | MIMAT0002473 |
Stem-loop Details
Lagothrix lagotricha
Pan paniscus

Genomic Map