Assay Details
Gene Family ID: | MIPF0000313, mir-306 |
Mature miRNA Sequence: | GGGGGUCACUCUGUGCCUGUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-306-3p | MIMAT0000394 |
| Drosophila erecta | der-miR-306-3p | MIMAT0008495 |
| Drosophila sechellia | dse-miR-306-3p | MIMAT0008805 |
| Drosophila yakuba | dya-miR-306-3p | MIMAT0009069 |
Stem-loop Details
Drosophila melanogaster
Drosophila erecta
Drosophila sechellia
Drosophila yakuba
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-306* (v16)
Drosophila erecta: der-miR-306* (v18)
Drosophila sechellia: dse-miR-306* (v18)
Drosophila yakuba: dya-miR-306* (v18)
Drosophila erecta: der-miR-306* (v18)
Drosophila sechellia: dse-miR-306* (v18)
Drosophila yakuba: dya-miR-306* (v18)

Genomic Map