Assay Details
Gene Family ID: | MIPF0001755, mir-462 |
Mature miRNA Sequence: | UAACGGAACCCAUAAUGCAGCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Zebrafish | dre-miR-462 | MIMAT0001855 |
Stem-loop Details
Zebrafish

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | dre-miR-462 |
---|---|
miRBase Accession Number: | MI0002028 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UAACGGAACCCAUAAUGCAGCU |
Species: | Zebrafish |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001755, mir-462 |
Mature miRNA Sequence: | UAACGGAACCCAUAAUGCAGCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Zebrafish | dre-miR-462 | MIMAT0001855 |