Assay Details
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | CAAAGUGCUCAUAGUGCAGGUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-20b-5p | MIMAT0021863 |
| Chrysemys picta | cpi-miR-20b-5p | MIMAT0037649 |
Stem-loop Details
Anolis carolinensis
Chrysemys picta
More Information
Related Products
miRBase Alias:
Aeromonas caviae Ae398: aca-miR-20b (v18)

Genomic Map