Assay Details
Gene Family ID: | MIPF0000757, mir-571 |
Mature miRNA Sequence: | UGAGUUGGCCAUCUGAGUGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-571 | MIMAT0003236 |
| Pongo pygmaeus | ppy-miR-571 | MIMAT0016014 |
Stem-loop Details
Human
Pongo pygmaeus

Genomic Map