Assay Details
Mature miRNA Sequence: | UGUCUACACUCAUGUUCUAGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Physcomitrella patens | ppt-miR1048-3p | MIMAT0005164 |
Stem-loop Details
Physcomitrella patens

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ppt-miR1048-3p |
---|---|
miRBase Accession Number: | MI0006007 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGUCUACACUCAUGUUCUAGA |
Species: | Physcomitrella patens |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UGUCUACACUCAUGUUCUAGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Physcomitrella patens | ppt-miR1048-3p | MIMAT0005164 |