Assay Details
Gene Family ID: | MIPF0000439, mir-434 | 
Mature miRNA Sequence: | UUUGAACCAUCACUCGACUCCU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-434-3p | MIMAT0001422 | 
| Rat | rno-miR-434-3p | MIMAT0005315 | 
Stem-loop Details
Mouse
Rat

Genomic Map