Assay Details
Gene Family ID: | MIPF0000319, mir-491 |
Mature miRNA Sequence: | CUUAUGCAAGAUUCCCUUCUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dog | cfa-miR-491 | MIMAT0006601 |
Stem-loop Details
Dog

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | cfa-miR-491 |
---|---|
miRBase Accession Number: | MI0007997 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CUUAUGCAAGAUUCCCUUCUA |
Species: | Dog |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000319, mir-491 |
Mature miRNA Sequence: | CUUAUGCAAGAUUCCCUUCUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dog | cfa-miR-491 | MIMAT0006601 |