Assay Details
Mature miRNA Sequence: | AGAAUUCAUAGCAAUACGGACU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-1619 | MIMAT0007486 |
Stem-loop Details
Chicken

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | gga-miR-1619 |
---|---|
miRBase Accession Number: | MI0007348 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGAAUUCAUAGCAAUACGGACU |
Species: | Chicken |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | AGAAUUCAUAGCAAUACGGACU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-1619 | MIMAT0007486 |