Assay Details
Mature miRNA Sequence: | AGUGGUUUUCCGUUGCACAUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Lottia gigantea | lgi-miR-1988 | MIMAT0009729 |
Stem-loop Details
Lottia gigantea
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | lgi-miR-1988 |
|---|---|
| miRBase Accession Number: | MI0010258 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGUGGUUUUCCGUUGCACAUGC |
| Species: | Lottia gigantea |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | AGUGGUUUUCCGUUGCACAUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Lottia gigantea | lgi-miR-1988 | MIMAT0009729 |