Assay Details
Gene Family ID: | MIPF0000360, mir-551 |
Mature miRNA Sequence: | GGCGACCCAUACUUGGUUUCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-551b | MIMAT0009348 |
Ophiophagus hannah | oha-miR-551b-3p | MIMAT0036943 |
Stem-loop Details
Bovine
Ophiophagus hannah
