Assay Details
Gene Family ID: | MIPF0000736, mir-2032 |
Mature miRNA Sequence: | AGGUAACUGUUGCAGCAUCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Nematostella vectensis | nve-miR-2032a-3p | MIMAT0009785 |
| Nematostella vectensis | nve-miR-2032b-3p | MIMAT0009787 |
Stem-loop Details
Nematostella vectensis
More Information
Related Products
miRBase Alias:
Nematostella vectensis: nve-miR-2032a* (v18)
Nematostella vectensis: nve-miR-2032b* (v18)
Nematostella vectensis: nve-miR-2032b* (v18)

Genomic Map