Assay Details
Gene Family ID: | MIPF0001195, MIR2112 |
Mature miRNA Sequence: | CUUUAUAUCCGCAUUUGCGCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR2112-3p | MIMAT0011155 |
| Arabidopsis lyrata | aly-miR2112-3p | MIMAT0017662 |
Stem-loop Details
Arabidopsis
Arabidopsis lyrata

Genomic Map