Assay Details
Mature miRNA Sequence: | UUGCAUAGUCUAUUAAGUUGGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Schmidtea mediterranea | sme-miR-2163-3p | MIMAT0011257 |
Stem-loop Details
Schmidtea mediterranea

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | sme-miR-2163-3p |
---|---|
miRBase Accession Number: | MI0010797 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUGCAUAGUCUAUUAAGUUGGU |
Species: | Schmidtea mediterranea |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UUGCAUAGUCUAUUAAGUUGGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Schmidtea mediterranea | sme-miR-2163-3p | MIMAT0011257 |