Assay Details
| Mature miRNA Sequence: | AAACAUCCGGACUAAGUUUAU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Schmidtea mediterranea | sme-miR-2204-3p | MIMAT0011399 | 
Stem-loop Details
Schmidtea mediterranea
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | sme-miR-2204-3p | 
|---|---|
| miRBase Accession Number: | MI0010937 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | AAACAUCCGGACUAAGUUUAU | 
| Species: | Schmidtea mediterranea | 
| Product Type: | mirVana® miRNA inhibitor | 
| Mature miRNA Sequence: | AAACAUCCGGACUAAGUUUAU | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Schmidtea mediterranea | sme-miR-2204-3p | MIMAT0011399 | 
 Back To Top
 Back To Top