Assay Details
Mature miRNA Sequence: | UCACACCUACAAUCCCUGGCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Zebrafish | dre-miR-2191 | MIMAT0011300 |
Stem-loop Details
Zebrafish

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | dre-miR-2191 |
---|---|
miRBase Accession Number: | MI0010842 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UCACACCUACAAUCCCUGGCA |
Species: | Zebrafish |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UCACACCUACAAUCCCUGGCA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Zebrafish | dre-miR-2191 | MIMAT0011300 |