Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GGAGAAAUUAUCCUUGCUGUGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Horse | eca-miR-539 | MIMAT0013167 |
Stem-loop Details
Horse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | eca-miR-539 |
---|---|
miRBase Accession Number: | MI0012910 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GGAGAAAUUAUCCUUGCUGUGU |
Species: | Horse |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GGAGAAAUUAUCCUUGCUGUGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Horse | eca-miR-539 | MIMAT0013167 |