Assay Details
Mature miRNA Sequence: | AUCGGUGAGCUAAAAAUAGAAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila melanogaster | dme-miR-2282-3p | MIMAT0011790 |
Stem-loop Details
Drosophila melanogaster

More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-2282 (v16)
