Assay Details
Gene Family ID: | MIPF0000836, mir-2808 |
Mature miRNA Sequence: | UCCGGCGAGAAAACUCAGUGGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2748 | MIMAT0012627 |
Stem-loop Details
Bombyx mori

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bmo-miR-2748 |
---|---|
miRBase Accession Number: | MI0012199 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UCCGGCGAGAAAACUCAGUGGG |
Species: | Bombyx mori |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000836, mir-2808 |
Mature miRNA Sequence: | UCCGGCGAGAAAACUCAGUGGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2748 | MIMAT0012627 |