Assay Details
Mature miRNA Sequence: | UACAGUGAUCAGGUUACGAUGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Columba livia | cli-miR-1329-5p | MIMAT0038672 |
| Taeniopygia guttata | tgu-miR-1329-5p | MIMAT0014449 |
Stem-loop Details
Columba livia
Taeniopygia guttata
More Information
Related Products
miRBase Alias:
Trabulsiella guamensis ATCC 49490: tgu-miR-1329 (v19)

Genomic Map