Assay Details
Gene Family ID: | MIPF0000856, mir-2543 |
Mature miRNA Sequence: | AUCCUAUCUGCUGUGGUUUAUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila pseudoobscura | dps-miR-2543a-5p | MIMAT0012344 |
Stem-loop Details
Drosophila pseudoobscura

More Information
Related Products
miRBase Alias:
Derxomyces pseudoschimicola: dps-miR-2543a* (v18)
