Assay Details
Gene Family ID: | MIPF0000829, mir-2774 |
Mature miRNA Sequence: | CUCGGUGGAAGACCUUCCCUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2774c | MIMAT0013759 |
Stem-loop Details
Bombyx mori

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bmo-miR-2774c |
---|---|
miRBase Accession Number: | MI0012450 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CUCGGUGGAAGACCUUCCCUU |
Species: | Bombyx mori |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000829, mir-2774 |
Mature miRNA Sequence: | CUCGGUGGAAGACCUUCCCUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2774c | MIMAT0013759 |