Assay Details
Gene Family ID: | MIPF0000980, mir-2513 |
Mature miRNA Sequence: | UAAAGCAACUUCAGACCAUAGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila pseudoobscura | dps-miR-2513a | MIMAT0012298 |
Stem-loop Details
Drosophila pseudoobscura
