Assay Details
Mature miRNA Sequence: | AAUGGUAAGAGAUUGAAGUGACU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pristionchus pacificus | ppc-miR-2270 | MIMAT0011725 |
Stem-loop Details
Pristionchus pacificus

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ppc-miR-2270 |
---|---|
miRBase Accession Number: | MI0011230 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AAUGGUAAGAGAUUGAAGUGACU |
Species: | Pristionchus pacificus |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | AAUGGUAAGAGAUUGAAGUGACU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pristionchus pacificus | ppc-miR-2270 | MIMAT0011725 |