Assay Details
Gene Family ID: | MIPF0000612, mir-564 |
Mature miRNA Sequence: | AGGCACGGUGGCAGCAGGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pongo pygmaeus | ppy-miR-564 | MIMAT0016009 |
Stem-loop Details
Pongo pygmaeus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ppy-miR-564 |
|---|---|
| miRBase Accession Number: | MI0015056 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGCACGGUGGCAGCAGGC |
| Species: | Pongo pygmaeus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000612, mir-564 |
Mature miRNA Sequence: | AGGCACGGUGGCAGCAGGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pongo pygmaeus | ppy-miR-564 | MIMAT0016009 |