Assay Details
Mature miRNA Sequence: | CACAGCGCAUACAAUGUGGAUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-460a-3p | MIMAT0026646 |
Columba livia | cli-miR-460a-3p | MIMAT0038653 |
Taeniopygia guttata | tgu-miR-460a-3p | MIMAT0014658 |
Stem-loop Details
Chicken
Columba livia
Taeniopygia guttata

More Information
Related Products
miRBase Alias:
Trabulsiella guamensis ATCC 49490: tgu-miR-460a* (v18)
