Assay Details
Mature miRNA Sequence: | CGUGUCUGUGUGUAUGUUGCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila simulans | dsi-miR-2580 | MIMAT0012517 |
Stem-loop Details
Drosophila simulans

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | dsi-miR-2580 |
---|---|
miRBase Accession Number: | MI0011786 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CGUGUCUGUGUGUAUGUUGCAG |
Species: | Drosophila simulans |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | CGUGUCUGUGUGUAUGUUGCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila simulans | dsi-miR-2580 | MIMAT0012517 |